missing translation for 'onlineSavingsMsg'
Learn More

Acrylic acid, 98%, extra pure, stabilized

Product Code. 10657753 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
1 L
10 L
2.5 L
250mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10657753 10 L Plastic drum 10 L
10488720 250 mL Glass bottle 250mL
10163890 1 L Glass bottle 1 L
10022020 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Product Code. 10657753 Supplier Thermo Scientific Acros Supplier No. 164250100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

CAS: 79-10-7 | C3H4O2 | 72.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-10-7 , 150-76-5
Molecular Formula C3H4O2
Molecular Weight (g/mol) 72.06
MDL Number MFCD00004367
InChI Key NIXOWILDQLNWCW-UHFFFAOYSA-N
Synonym acrylic acid, 2-propenoic acid, propenoic acid, vinylformic acid, acroleic acid, propene acid, ethylenecarboxylic acid, polyacrylate, propenoate, carbomer
PubChem CID 6581
ChEBI CHEBI:18308
IUPAC Name prop-2-enoic acid
SMILES C=CC(=O)O

Specifications

CAS Min % 98
Melting Point 13.0°C
Density 1.0500g/mL
Boiling Point 139.0°C
Flash Point 48°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic drum
Linear Formula H2C=CHCO2H
Refractive Index 1.4192 to 1.4212
Quantity 10 L
Beilstein 02, 397
Merck Index 15, 121
Specific Gravity 1.05
Solubility Information Solubility in water: miscible. Other solubilities: miscible with most organic solvents,soluble in acetone
Stabilizer 180 to 220ppm MEHQ
Viscosity 1.3 mPa.s (20°C)
Formula Weight 72.06
Percent Purity 98%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material Acrylic acid, Stabilized
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Acrylic acid, stabilized
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.