missing translation for 'onlineSavingsMsg'
Learn More

Acrylamide, Ultrapure Bioreagent, J.T. Baker™

Product Code. 15508244 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15508244 500 g Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 15508244 Supplier Avantor J.T.Baker Supplier No. 408101

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

Specifications

CAS 79-06-1
Quantity 500 g
Flash Point 138.00°C
Packaging Poly Bottle
Molecular Formula C3H5NO
MDL Number MFCD00008032
InChI Key HRPVXLWXLXDGHG-UHFFFAOYSA-N
SMILES NC(=O)C=C
IUPAC Name prop-2-enamide
Molecular Weight (g/mol) 71.08
PubChem CID 6579
ChEBI CHEBI:28619
Formula Weight 71.08
Grade Ultrapure Bioreagent
Chemical Name or Material Acrylamide, Purified for Electrophoresis
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.