missing translation for 'onlineSavingsMsg'
Learn More

Acrylamide-d3 (1.0 mg/mL in Deionized water with 0.1% Formic Acid), TRC

Product Code. 30470651
Change view
Click to view available options
Quantity:
2 x 1 mL
Unit Size:
2mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30470651 2 x 1 mL 2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30470651 Supplier Toronto Research Chemicals Supplier No. TRCKIT97922X1ML
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 122775-19-3
Molecular Formula C3 2H3 H2 N O
Quantity 2 x 1 mL
Synonym 2-Propenamide-2,3,3-d3, Acrylamide-2,3,3 D3, Acrylamide-d3, Acrylamide-2,2,2-D3
InChI Formula InChI=1S/C3H5NO/c1-2-3(4)5/h2H,1H2,(H2,4,5)/i1D2,2D
SMILES [2H]C(=C([2H])C(=O)N)[2H]
IUPAC Name 2,3,3-trideuterioprop-2-enamide
Molecular Weight (g/mol) 74.1
Formula Weight 74.0559
Concentration or Composition (by Analyte or Components) Acrylamide-d3 1 mg/mL
Chemical Name or Material Acrylamide-d3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.