missing translation for 'onlineSavingsMsg'
Learn More

Acrylamide: Bis-Acrylamide 37.5:1 (40% Solution/Electrophoresis), Fisher BioReagents

Product Code. 30117569
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30117569 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 30117569 Supplier Fisher BioReagents Supplier No. BP14101
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
This Acrylamide Solution is premixed for 2.6% cross-linking and is suitable for separation of high-molecular-weight proteins.
TRUSTED_SUSTAINABILITY

Specifications

Physical Form Liquid
CAS 79-06-1, 110-26-9, 7732-18-5
Conductivity 10μmhos max.
Color Colorless
Quantity 1 L
Packaging Polypropylene Bottle
Molecular Formula C3H5NO
MDL Number MFCD00008032
InChI Key HRPVXLWXLXDGHG-UHFFFAOYSA-N
SMILES NC(=O)C=C
IUPAC Name prop-2-enamide
Molecular Weight (g/mol) 71.08
PubChem CID 6579
ChEBI CHEBI:28619
Grade DNase-, RNase- and Protease-Free
Chemical Name or Material Acrylamide: Bis-Acrylamide 37.5:1, 40% Solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.