missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Acrylamide, 99.5%, Molecular Biology Grade

Product Code. 15487439
Change view
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15487439 1 kg 1kg
15477439 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 15487439 Supplier Thermo Scientific Chemicals Supplier No. J66184.A1
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

CAS: 79-06-1 | C3H5NO | 71.08 g/mol

Acrylamide solution is a commonly used matrix for electrophoretic separation of nucleic acids and proteins. Also used for preparing slab gel gradients for PAGE.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acrylamide solution is a commonly used matrix for electrophoretic separation of nucleic acids and proteins. Also used for preparing slab gel gradients for PAGE.

Solubility
Partly soluble in water.

Notes
Air and Light Sensitive, store in dark and away form light. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-06-1
Molecular Formula C3H5NO
Molecular Weight (g/mol) 71.08
MDL Number MFCD00008032
InChI Key HRPVXLWXLXDGHG-UHFFFAOYSA-N
Synonym acrylamide, 2-propenamide, propenamide, acrylic amide, akrylamid, ethylenecarboxamide, vinyl amide, acrylic acid amide, 2-propeneamide, propeneamide
PubChem CID 6579
ChEBI CHEBI:28619
IUPAC Name prop-2-enamide
SMILES NC(=O)C=C

Specifications

Melting Point 82°C to 85°C
Color White
Density 1.322 g/mL
Boiling Point 125°C (25 mmHg)
Flash Point 138°C (280°F)
Odor Odorless
Linear Formula CH2=CHCONH2
Quantity 1 kg
UN Number UN2074
Beilstein 605349
Sensitivity Air and light sensitive
Merck Index 14,129
Solubility Information Partly soluble in water.
Formula Weight 71.08
Percent Purity 99.5%
Grade Molecular Biology
Physical Form Powder
Chemical Name or Material Acrylamide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.