missing translation for 'onlineSavingsMsg'
Learn More

Acrylamide (1.0 mg/mL in Deionized water with 0.1% Formic Acid), TRC

Product Code. 30010117
Change view
Click to view available options
Quantity:
2 x 1 mL
Unit Size:
2mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010117 2 x 1 mL 2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 30010117 Supplier Toronto Research Chemicals Supplier No. TRCKIT97902X1ML
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 79-06-1
Molecular Formula C3 H5 N O
Quantity 2 x 1 mL
Synonym Acrylamide (8CI), Acrylic amide, Bio-Acrylamide 50, DM 206, Ethylenecarboxamide, NSC 7785, Propenamide, Vinyl amide
InChI Formula InChI=1S/C3H5NO/c1-2-3(4)5/h2H,1H2,(H2,4,5)
SMILES NC(=O)C=C
IUPAC Name prop-2-enamide
Molecular Weight (g/mol) 71.08
Formula Weight 71.0371
Concentration or Composition (by Analyte or Components) Acrylamide 1 mg/mL
Chemical Name or Material Acrylamide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.