missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acrosin Antibody (ACR-2), PE/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-419PECY7
This item is not returnable.
View return policy
Description
Acrosin Monoclonal specifically detects Acrosin in Porcine, Bovine (Negative), Canine (Negative), Human (Negative) samples. It is validated for Flow Cytometry.
Specifications
| Acrosin | |
| Monoclonal | |
| PE-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| ACR | |
| Acid extracts of boar spermatozoa were subjected to hydrophobic chromatography and the pooled fraction with reactivity to N-alpha benzoylarginine-4-nitroanilide was used for immunization. | |
| 0.1 mL | |
| Primary | |
| The antibody ACR-2 reacts with various forms of porcine acrosin (55, 53, 45 and 35 kDa), a typical serine proteinase with trypsin-like specificity. Acrosin is stored in the acrosome of undamaged spermatozoa. | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| ACR-2 | |
| Flow Cytometry | |
| acrosin, ACRS, preproacrosin, proacrosin | |
| Mouse | |
| Protein A purified | |
| Signal Transduction | |
| 49.0 | |
| Pig, Bovine (Negative), Canine (Negative), Human (Negative) | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction