missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acrosin Antibody (ACR-2), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-419AF700
This item is not returnable.
View return policy
Description
Acrosin Monoclonal specifically detects Acrosin in Porcine, Bovine (Negative), Canine (Negative), Human (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| Acrosin | |
| Monoclonal | |
| Alexa Fluor 700 | |
| acrosin, ACRS, preproacrosin, proacrosin | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| The antibody ACR-2 reacts with various forms of porcine acrosin (55, 53, 45 and 35 kDa), a typical serine proteinase with trypsin-like specificity. Acrosin is stored in the acrosome of undamaged spermatozoa. | |
| Store at 4C in the dark. |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| ACR-2 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| ACR | |
| Acid extracts of boar spermatozoa were subjected to hydrophobic chromatography and the pooled fraction with reactivity to N-alpha benzoylarginine-4-nitroanilide was used for immunization. | |
| 0.1 mL | |
| Signal Transduction | |
| 49 | |
| Pig, Bovine (Negative), Canine (Negative), Human (Negative) | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction