Learn More
Cytiva AcroPrep™ Advance 96-well Filter Plates for Ultrafiltration
Product Code. 17184621
Change view
Click to view available options
Volume (Metric):
350 μL
1 mL
MWCO:
10 kDa
100 kDa
3 kDa
30 kDa
Unit Size:
Pack of 10
Pack of 5
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) MWCO unitSize
17184621 1 mL 3 kDa Pack of 5
17194601 350 μL 30 kDa Pack of 10
17184601 350 μL 10 kDa Pack of 10
17104611 350 μL 100 kDa Pack of 10
17174601 350 μL 3 kDa Pack of 10
17104631 1 mL 30 kDa Pack of 5
17194621 1 mL 10 kDa Pack of 5
17114631 1 mL 100 kDa Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 17184621 Supplier Cytiva Supplier No. 8163
Only null left
Add to basket
Add to basket
Optimized for a variety of applications requiring larger per well volume processing.
  • Plates are constructed of chemically resistant, biologically-inert polypropylene
  • This product is not approved for use in medical, clinical, surgical or other patient applications
TRUSTED_SUSTAINABILITY
Specifications
Material (Membrane) Omega (Modified PES)
MWCO 3 kDa
Sterility Non-sterile
Housing Material Polypropylene Housing
Quantity 5/Pk.
Volume (Metric) 1 mL