Learn More
Cytiva AcroPrep™ Advance 96-well Filter Plates for Ultrafiltration
Description
- Plates are constructed of chemically resistant, biologically-inert polypropylene
- This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
Specifications
| Material (Membrane) | Omega (Modified PES) |
| MWCO | 100 kDa |
| Sterility | Non-sterile |
| Housing Material | Polypropylene Housing |
| Quantity | 5/Pk. |
| Volume (Metric) | 1 mL |