Learn More
Cytiva AcroPrep™ Advance 96-well Filter Plates for Solvent Filtration
Description
- Plates are constructed of chemically resistant, biologically-inert polypropylene
- This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
Specifications
| Material (Membrane) | PTFE |
| Pore Size | 0.2 μm |
| Sterility | Non-sterile |
| For Use With (Application) | Solvent Filtration |
| Quantity | 10/Pk. |
| Volume (Metric) | 350 μL |