Learn More
Cytiva AcroPrep™ Advance 96-well Filter Plates for Lysate Clearance
Description
- Plates are constructed of chemically resistant, biologically-inert polypropylene
- This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
Specifications
| Material (Membrane) | Borosilicate Glass w/o Binder, Supor (PES) |
| Pore Size | 3.0/0.2 μm |
| Sterility | Non-sterile |
| Housing Material | Polypropylene Housing |
| Quantity | 5/Pk. |
| Volume (Metric) | 1 mL |