missing translation for 'onlineSavingsMsg'
Learn More

Cytiva (formerly Pall Lab) AcroPrep™ Advance 96-well Filter Plates for Aqueous Filtration

Optimized for a variety of applications requiring larger per well volume processing.

Brand:  Cytiva (formerly Pall Lab) 8231

Additional Details : Weight : 1.94750kg

 View more versions of this product

Product Code. 17164631

  • £155.00 / Pack of 5
Estimated Shipment: 10-06-2024
to see stock.
Add to basket

Description

Description

  • Plates are constructed of chemically resistant, biologically-inert polypropylene
  • This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications

Specifications

Borosilicate Glass w/o Binder, Supor (PES)
Non-sterile
5/Pk.
1.0 μm
Polypropylene Housing
2 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers