missing translation for 'onlineSavingsMsg'
Learn More

Cytiva AcroPrep™ 24-well Filter Plates
GREENER_CHOICE

Product Code. 17184651
Change view
Click to view available options
Quantity:
2/Pk.
8/Pk.
Membrane Type:
Omega
PP/PE
Supor
Pore Size:
0.1 μm
0.45 μm
0.65/0.2 μm
0.65/0.2 μm with Depth Filter
0.8 μm
1.2 μm
30 to 40 μm
5 μm
Unit Size:
Pack of 2
Pack of 8
MWCO:
1 kDa
10 kDa
100 kDa
3 kDa
30 kDa
50 kDa
28 product options available for selection
Product selection table with 28 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Membrane Type Pore Size unitSize MWCO
17184651 8/Pk. Supor 0.65/0.2 μm with Depth Filter Pack of 8 -
17164651 2/Pk. Supor 0.65/0.2 μm with Depth Filter Pack of 2 -
17174651 2/Pk. Supor 0.65/0.2 μm Pack of 2 -
17194651 8/Pk. Supor 0.65/0.2 μm Pack of 8 -
17309481 2/Pk. Supor 0.1 μm Pack of 2 -
17399471 8/Pk. Supor 0.1 μm Pack of 8 -
17329481 2/Pk. Supor 0.45 μm Pack of 2 -
17319481 8/Pk. Supor 0.45 μm Pack of 8 -
17349481 2/Pk. Supor 0.8 μm Pack of 2 -
17339481 8/Pk. Supor 0.8 μm Pack of 8 -
17369481 2/Pk. Supor 1.2 μm Pack of 2 -
17359481 8/Pk. Supor 1.2 μm Pack of 8 -
17389481 2/Pk. Supor 5 μm Pack of 2 -
17379481 8/Pk. Supor 5 μm Pack of 8 -
17309491 2/Pk. Omega - Pack of 2 1 kDa
17399481 8/Pk. Omega - Pack of 8 1 kDa
17329491 2/Pk. Omega - Pack of 2 3 kDa
17319491 8/Pk. Omega - Pack of 8 3 kDa
17349491 2/Pk. Omega - Pack of 2 10 kDa
17339491 8/Pk. Omega - Pack of 8 10 kDa
17369491 2/Pk. Omega - Pack of 2 30 kDa
17359491 8/Pk. Omega - Pack of 8 30 kDa
17389491 2/Pk. Omega - Pack of 2 50 kDa
17379491 8/Pk. Omega - Pack of 8 50 kDa
17309501 2/Pk. Omega - Pack of 2 100 kDa
17399491 8/Pk. Omega - Pack of 8 100 kDa
17329501 2/Pk. PP/PE 30 to 40 μm Pack of 2 -
17319501 8/Pk. PP/PE 30 to 40 μm Pack of 8 -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 28 options available.
28 options
This item is not returnable. View return policy
Product Code. 17184651 Supplier Cytiva Supplier No. 97026
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

A one-and-done solution for filtration in a 24-well format: the AcroPrep™ 24-well filter plate range. For applications including protein purification, sterile filtration, general aqueous filtration, desalting, diafiltration, concentration, and fractionation of protein and nucleic acid samples.

Utilizing proprietary high-performance multi-layer filter membranes, plates offer time savings strong performance claims and streamlined workflow improvements. Available in 4 different types of membranes. Each plate comes with a V-shaped bottom collection plate and lid. Automation friendly. Vacuum compatible. Suitable for centrifuge.

 

Plates with Asymmetric Supor™ EKV Membrane:

A 0.65/0.2 μm asymmetric Supor EKV PES membrane, available with and without a dual-layer depth prefilter. The depth prefilter allows clarification of large cellular debris prior to filtration through the 0.65/0.2 μm EKV Supor membrane. Dual-layer clarification plate has reliable recovery of > 95% of extra cellular proteins. Single-layer AcroPrep 24-well filter plate contains 0.65/0.2 μm Supor EKV membrane, and is designed for applications where a sterile filtrate is required.

 

Plates with Supor Membrane:

The AcroPrep Supor plates have a variety of uses, in applications such as resin packing for sample preparation, viral filtration, or genome mapping, depending on the pore size chosen. Available with 0.1, 0.45, 0.8, 1.2 and 5 μm pore size ratings, these versatile filter plates can be confidently incorporated into a number of laboratory 24-well format workflows. The Supor polyethersulfone membrane enables fast filtration, has low protein binding properties and offers extensive drug and chemical compatibility. Each plate comes with a V-shaped bottom collection plate and lid Individually bagged and gamma irradiated. Automation friendly vacuum compatible suitable for centrifuge.

 

Plates with Omega Membrane:

The AcroPrep Omega plate for desalting, diafiltration, concentration or fractionation of protein and nucleic acid samples saves time in a single device and workflow step. The Omega ultrafiltration membrane is a polyethersulfone membrane specifically modified to minimize protein and nucleic acid binding. The Omega membrane works on the premise of size exclusion, where samples are fractionated by molecular weight. Larger molecules can be separated from smaller molecules by selecting the appropriate molecular weight cut-off (MWCO) and filtering the sample through these plates by either centrifugation, vacuum, or positive pressure, typically resulting in ≥ 90% recovery of target molecules.

 

Plates with PE/PP Membrane: 

The PE/PP plate offers a large pore size (30 to 40 μm) which is ideal for plant applications or as a cell strainer.

TRUSTED_SUSTAINABILITY

Specifications

Material Polypropylene
Pore Size 0.65/0.2 μm with Depth Filter
Sterility Sterile
Filtration Area 1.6 cm2
Membrane Type Supor
Quantity 8/Pk.
Type Filter Plate with Membrane
Volume (Metric) 7 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.