Learn More
Cytiva (formerly Pall Lab) AcroPak™ 800 Capsules with Supor™ EKV Membrane
AcroPak capsules designed for research applications only.
Brand: Cytiva (formerly Pall Lab) 12463
Additional Details : Weight : 0.26800kg
Description
- For buffers, tissue culture media, and other biological fluids
- Newer technology recommended for simplified scale-up
- High filtration area and compact size are ideal for upscale trials
Specifications
Hydrophilic PES | |
Sterile | |
1 to 1.5 in. Sanitary Flange |
0.2 μm | |
750 cm2 | |
1/Ea. |