Learn More
Cytiva AcroPak™ 300 Capsule with PTFE Membrane
Product Code. 17124221
Click to view available options
No. per Pack:
100/Pk.
3/Pk.
Unit Size:
Pack of 100
Pack of 3
Product Code. 17124221
Brand: Cytiva 12085
AcroPak 300 Capsule offers broad chemical compatibility with PTFE membrane and polypropylene housing.
  • High air flow rate venting applications
  • Offers broad chemical compatibility with PTFE membrane and polypropylene housing 100% integrity-tested to provide the high level of confidence you require
  • This product is not approved for use in medical, clinical, surgical or other patient applications
TRUSTED_SUSTAINABILITY
Specifications
Material (Membrane) PTFE
Pore Size 0.2 μm
Sterility Non-sterile
Filtration Area 280 cm2
No. per Pack 100/Pk.
Outlet Style Stepped Hose Barb