Learn More
Thermo Scientific™ AcroMetrix™ HIV-1/HCV/HBV Panels (RUO)
Product Code. 17633545 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
AcroMetrix HBV Panel
Level:
50, 500, 5000, 50000, 500000, 5000000, 50000000 IU/mL
Unit Size:
Each
Product Code. Description Level Unit Size
17633545 AcroMetrix HBV Panel 50, 500, 5000, 50000, 500000, 5000000, 50000000 IU/mL Each
1 options
Product Code. 17633545 Supplier Thermo Scientific™ Supplier No. 950150
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Assess the performance of molecular test procedures for HIV-1, HCV and HBV nucleic acids with these panels, which are for Research Use Only.
Assess the performance of molecular test procedures, including the extraction step, for the quantitative and qualitative determination of HIV-1, HCV and HBV nucleic acids with the Thermo Scientific™ AcroMetrix™ HIV-1/HCV/HBV Panels (RUO). These panels can be used for training and proficiency testing of laboratory personnel, for lot-to-lot comparison of reagent test kits and to evaluate and compare intralaboratory and interlaboratory assessments. The panels are for Research Use Only and are not for use in diagnostic procedures.
TRUSTED_SUSTAINABILITY
Specifications
Description AcroMetrix HBV Panel
Detectable Analytes HBV, HCV, HBV
Type HBV Panel
Level 50, 500, 5000, 50000, 500000, 5000000, 50000000 IU/mL
Packaging 7 x 1.2mL
Quantity 7 x 1.2 mL