missing translation for 'onlineSavingsMsg'
Learn More

Acrolein diethyl acetal, 96%

Product Code. 11446586
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11446586 10 g 10g
11456586 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11446586 Supplier Thermo Scientific Alfa Aesar Supplier No. B24821.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3054-95-3 | C7H14O2 | 130.187 g/mol

Acrolein diethyl acetal is a reagent used for the synthesis of a variety of compounds, including cinnamaldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acrolein diethyl acetal is a reagent used for the synthesis of a variety of compounds, including cinnamaldehydes.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3054-95-3
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.187
MDL Number MFCD00009239
InChI Key MCIPQLOKVXSHTD-UHFFFAOYSA-N
Synonym acrolein diethyl acetal, acrolein diethylacetal, acrolein acetal, 3,3-diethoxy-1-propene, 1-propene, 3,3-diethoxy, propenal diethyl acetal, 3,3-diethoxypropene, 1,1-diethoxy-2-propene, acrolein, diethyl acetal, acrylaldehyde diethyl acetal
PubChem CID 62477
IUPAC Name 3,3-diethoxyprop-1-ene
SMILES CCOC(C=C)OCC

Specifications

Density 0.852
Boiling Point 124°C to 125°C
Flash Point 13°C (55°F)
Refractive Index 1.401
Quantity 10 g
UN Number UN2374
Beilstein 1701567
Solubility Information Slightly soluble in water.
Formula Weight 130.19
Percent Purity 96%
Chemical Name or Material Acrolein diethyl acetal
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.