missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pall Acrodisc™ Unit with Mustang Q Membrane
Disposable chromatography units with high binding capacities and fast flow rates.
Brand: Pall MSTG25Q6
Additional Details : Weight : 0.39900kg
Description
- Mustang Q membrane is a strong anion exchanger that effectively binds plasmid DNA, negatively-charged proteins, and viral particles
- For research use only
Specifications
Mustang Q | |
Non-sterile | |
Blister Pack |
0.8 μm | |
10/Pk. | |
25 mm |