missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Acrodisc™ Syringe Filters with Supor™ Membrane, Sterile

Product Code. 17154361
Change view
Click to view available options
Pore Size:
0.1 μm
0.2 μm
0.45 μm
0.8 μm
1.2 μm
5 μm
Diameter (Metric):
13 mm
25 mm
32 mm
Unit Size:
Pack of 50
Pack of 75
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size Diameter (Metric) unitSize
17154361 0.2 μm 13 mm Pack of 75
17184361 0.1 μm 25 mm Pack of 50
17114381 0.1 μm 32 mm Pack of 50
17194361 0.2 μm 25 mm Pack of 50
17124381 0.2 μm 32 mm Pack of 50
17164361 0.45 μm 13 mm Pack of 75
17104371 0.45 μm 25 mm Pack of 50
17144381 0.45 μm 32 mm Pack of 50
17174361 0.8 μm 13 mm Pack of 75
17114371 0.8 μm 25 mm Pack of 50
17164381 1.2 μm 32 mm Pack of 50
17104381 5 μm 32 mm Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Product Code. 17154361 Supplier Cytiva Supplier No. 4602
Only null left
Add to basket
Edge
Add to basket

Sterile Acrodisc syringe filters featuring Supor™ membrane for high throughputs, fast filtration, and low protein binding. Available in 13 mm, 25 mm and 32 mm sizes, with pore sizes ranging from 0.1 μm to 5.0 μm.

The syringe filter is the laboratory workhorse that protects sample or culture quality and sterility.

The Acrodisc product line includes high-performance dual-membrane and combination filter technology which gives improved throughput and higher debris clean up in a single syringe filter. These filters help provide enough capacity to filter an entire sample in one go.

  • Gamma-irradiated individual blister packs ensure sterility
  • Ergonomic blister packs assure sterile storage, handling, and connections
  • Superior membrane performance achieves high filtration flow and throughput
  • Integrated dual-layer pre-filter for high dirt-holding capacity and increased throughput
  • Low protein binding ensures high protein recovery

Not recommended for use with some ketones.

This product is not approved for use in medical, clinical, surgical or other patient applications.

TRUSTED_SUSTAINABILITY

Specifications

Material (Membrane) PES
Pore Size 0.2 μm
Sterility Sterile
Filtration Area 1 cm2
Quantity 75/Pk.
Diameter (Metric) 13 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.