missing translation for 'onlineSavingsMsg'
Learn More

Cytiva (formerly Pall Lab) Acrodisc™ Syringe Filters with Glass Fiber Filter

Available with GxF multi-layered glass fiber prefilter, providing two to four times the throughput of standard glass fiber prefilter devices, and allowing quick and easy filtration of your most difficult-to-filter samples.

Brand:  Cytiva (formerly Pall Lab) 4529

2577.24 GBP valid until 2024-05-06
Use promo code "21711" to get your promotional price.



Additional Details : Weight : 3.21900kg

 View more versions of this product

Product Code. 17174341

  • £3400.00 / Pack of 1000
Estimated Shipment: 23-05-2024
to see stock.
Add to basket

Description

Description

  • For research use only
Specifications

Specifications

Borosilicate Glass Fiber
Non-sterile
1000/Pk.
1 μm
Polypropylene
25 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers