missing translation for 'onlineSavingsMsg'
Learn More

Acrizanib, MedChemExpress

Product Code. 16465186
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16465186 100 mg 100mg
16460587 50 mg 50mg
16456977 5 mg 5mg
16444258 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16465186 Supplier Medchem Express Supplier No. HY109020
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Acrizanib (LHA510) is a VEGFR-2 inhibitor, with an IC50 of 17.4 nM for BaF3-VEGFR-2.

Acrizanib (LHA510) is a VEGFR-2 inhibitor, with an IC50 of 17.4 nM for BaF3-VEGFR-2.

Specifications

Chemical Name or Material Acrizanib
CAS 1229453-99-9
Color White
Purity Grade Notes Research
Molecular Formula C20H18F3N7O2
Quantity 100 mg
Synonym LHA510
Solubility Information DMSO : 41.67 mg/mL (93.56 mM; Need ultrasonic)
SMILES O=C(N1C2=CC=C(OC3=NC=NC(CNC)=C3)C=C2C=C1)NC4=NN(C)C(C(F)(F)F)=C4
Molecular Weight (g/mol) 445.4
Formula Weight 445.4
Percent Purity 98.23%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.