missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 260.0°C |
| Color | Brown-Red to Orange-Red |
| Loss on Drying | 7% max. (105°C, 1 hr.) |
| Infrared Spectrum | Authentic |
| Packaging | Glass bottle |
| Quantity | 25 g |
| Beilstein | 22,I,65 |
| Merck Index | 15,115 |
| Solubility Information | Solubility in water: 250-300g/L (20°C). Other solubilities: soluble in alcohol,practically insoluble in ether and chloroform |
| Physical Form | Powder |
| Show More |
Safety and Handling
Product Identifier
- Acriflavine hydrochloride
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 1
- H318-Causes serious eye damage.
- H411-Toxic to aquatic life with long lasting effects.
- H302-Harmful if swallowed.
- P264-Wash face, hands and any thoroughly after handling.
- P273-Avoid release to the environment.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?