missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Acriflavine

Product Code. 16434386
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16434386 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16434386 Supplier MedChemExpress Supplier No. HY100575

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Acriflavine is a fluorescent dye for labeling high molecular weight RNA. It is also a topical antiseptic.

Specifications

CAS 8048-52-0
Solubility H2O : ≥ 25 mg/mL (96.25 mM)
Synonym Acriflavinium chloride 3,6-Acridinediamine mix
Content And Storage 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
For Use With (Application) Cancer-programmed cell death
Quantity 100 mg
Molecular Weight (g/mol) 259.73 g/mol
Product Type Dye Reagents
Purity or Quality Grade 98.09%
Shipping Condition Room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.