missing translation for 'onlineSavingsMsg'
Learn More

Acriflavin-Biotin Conjugate (~90%), TRC

Product Code. 30309056
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30309056 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30309056 Supplier Toronto Research Chemicals Supplier No. TRCA1911001MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acriflavin-Biotin Conjugate
CAS 1041387-90-9
Molecular Formula C25 H31 N7 O2 S
Quantity 1 mg
Synonym Acriflavin-Biotin Conjugate
InChI Formula InChI=1S/C25H31N7O2S/c26-14-5-7-18-16(11-14)23(17-12-15(27)6-8-19(17)30-18)29-10-9-28-22(33)4-2-1-3-21-24-20(13-35-21)31-25(34)32-24/h5-8,11-12,20-21,24H,1-4,9-10,13,26-27H2,(H,28,33)(H,29,30)(H2,31,32,34)/t20-,21-,24-/m0/s1
SMILES Nc1ccc2nc3ccc(N)cc3c(NCCNC(=O)CCCC[C@@H]4SC[C@@H]5NC(=O)N[C@H]45)c2c1
IUPAC Name 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]-N-[2-[(2,7-diaminoacridin-9-yl)amino]ethyl]pentanamide
Molecular Weight (g/mol) 493.62
Formula Weight 493.226
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.