missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Acridine Orange, pure, ca. 55% dye content

Product Code. 10471681 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10471681 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10471681 Supplier Thermo Scientific Chemicals Supplier No. 300911000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10127-02-3 | C17H20Cl3N3Zn | 438.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 165.0°C
CAS 10127-02-3
Dye Content 50% min.
Molecular Formula C17H20Cl3N3Zn
MDL Number MFCD00081043,MFCD00081043,MFCD00081043
Synonym Basic Orange 14, 3,6-Bis(dimethylamino)acridine zinc chloride double salt, C.I. 46005
Solubility Information Solubility in water: soluble.
InChI Key VADJQOXWNSPOQA-UHFFFAOYSA-L
Infrared Spectrum Authentic
SMILES [H+].[Cl-].[Cl-].[Cl-].[Zn++].CN(C)C1=CC2=NC3=CC(=CC=C3C=C2C=C1)N(C)C
IUPAC Name zinc;hydron;3-N,3-N,6-N,6-N-tetramethylacridine-3,6-diamine;trichloride
Molecular Weight (g/mol) 438.10
PubChem CID 71774334
Formula Weight 369.94
Grade Pure
Quantity 100 g
Chemical Name or Material Acridine Orange
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.