missing translation for 'onlineSavingsMsg'
Learn More

Acridine Orange Hemizinc Salt, Fisher BioReagents™

Product Code. 10667433
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10667433 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10667433 Supplier Fisher BioReagents Supplier No. BP11610
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Acridine Orange is suitable for microscopy and as a specific stain for DNA and RNA.
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 165°C
CAS 10127-02-3
Extinction Coefficient 1100g(-1) cm(-1) min.
Molecular Formula C17H20Cl3N3Zn
MDL Number MFCD00081043,MFCD00081043,MFCD00081043
Identification Pass Test
InChI Key VADJQOXWNSPOQA-UHFFFAOYSA-L
SMILES [H+].[Cl-].[Cl-].[Cl-].[Zn++].CN(C)C1=CC2=NC3=CC(=CC=C3C=C2C=C1)N(C)C
IUPAC Name zinc;hydron;3-N,3-N,6-N,6-N-tetramethylacridine-3,6-diamine;trichloride
Molecular Weight (g/mol) 438.10
PubChem CID 71774334
Lambda Max 488 to 492nm
Formula Weight 369.94
Loss on Drying 5% max.
Packaging Amber Glass
Color Index No. 46005
Color Orange
Quantity 10 g
Physical Form Solid
Chemical Name or Material Acridine Orange Hemizinc Salt
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.