missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACRBP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101205
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ACRBP.
The protein encoded by this gene is similar to proacrosin binding protein sp32 precursor found in mouse, guinea pig, and pig. This protein is located in the sperm acrosome and is thought to function as a binding protein to proacrosin for packaging and condensation of the acrosin zymogen in the acrosomal matrix. This protein is a member of the cancer/testis family of antigens and it is found to be immunogenic. In normal tissues, this mRNA is expressed only in testis, whereas it is detected in a range of different tumor types such as bladder, breast, lung, liver, and colon. [provided by RefSeq, Jul 2008].
Specifications
| ACRBP | |
| Polyclonal | |
| Unconjugated | |
| ACRBP | |
| ACRBP; acrbp {ECO:0000312; acrosin binding protein; acrosin-binding protein; Cancer/testis antigen 23; cancer/testis antigen OY-TES-1; CT23; EMBL:AAH79212.1}; OY-TES-1; proacrosin binding protein; proacrosin binding protein sp32; proacrosin-binding protein sp32; SP32; testicular tissue protein Li 10 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 500316, 84519 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q6AY33, Q8NEB7 | |
| ACRBP | |
| A synthesized peptide derived from human ACRBP(Accession Q8NEB7), corresponding to amino acid residues L186-Q236. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction