missing translation for 'onlineSavingsMsg'
Learn More

Waters Corporation ACQUITY UPLC Protein SEC Column and BEH450 SEC Protein Standard Mix

Product Code. p-7236646
Encompass
Click to view available options
Length (Metric):
30 mm
150 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17122622

Brand: Waters Corporation 176002996

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ACQUITY UPLC BEH450 SEC Column based upon the highly successful BEH particle technology.

  • Based upon the highly successful BEH particle technology, the ACQUITY UPLC BEH450 SEC column is fully optimized for the analysis of proteins and their aggregates with molecular weights ranging from 100,000 to 1,500,000 Daltons, the total system solution will routinely deliver fast, accurate aggregate determinations up to 10 times quicker that traditional HPLC-based SEC methods.

Specifications

Diameter (Metric) 4.6 mm
Length (Metric) 150 mm
Particle Shape Spherical
Particle Size 2.5 μm
Pore Size 450 Å
Quantity 1/Ea.
USP Type L33
Endcapped No
Type Column
For Use With (Application) Protein
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.