missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Waters Corporation ACQUITY UPLC BEH200 SEC Column
Click to view available options
Length (Metric):
30 mm
150 mm
Unit Size:
Each
Description
- Fully optimized for the analysis of proteins and their aggregates with molecular weights ranging from 10,000 to 450,000 Daltons, the total system solution will routinely deliver fast, accurate aggregate determinations up to 10 times quicker that traditional HPLC-based SEC methods.
Specifications
Specifications
| Diameter (Metric) | 4.6 mm |
| Length (Metric) | 150 mm |
| Particle Shape | Spherical |
| Particle Size | 1.7 μm |
| Pore Size | 200 Å |
| Quantity | 1/Ea. |
| USP Type | L33 |
| Endcapped | No |
| Type | Column |
| For Use With (Application) | Protein |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction