missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ACPL2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA553567

Product Code. 15781501

  • £467.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: STPKNGMSSK SRKRIMPDPV TEPPVTDPVY EALLYCNIPS VAERSMEGHA PHHFKLVSVH VFIRHGDRYP LYVIPKTKRP EIDCTLVANR KPYHPKLEAF ISHMSKGSGA SFESPLNSLP LYPNHP Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 90%.

Responsible for the 2-O-dephosphorylation of xylose in the glycosaminoglycan-protein linkage region of proteoglycans thereby regulating the amount of mature glycosaminoglycan (GAG) chains. Sulfated glycosaminoglycans (GAGs), including heparan sulfate and chondroitin sulfate, are synthesized on the so-called common GAG-protein linkage region (GlcUAbeta1-3Galbeta1-3Galbeta1-4Xylbeta1-O-Ser) of core proteins, which is formed by the stepwise addition of monosaccharide residues by the respective specific glycosyltransferases. Xylose 2-O-dephosphorylation during completion of linkage region formation is a prerequisite for the initiation and efficient elongation of the repeating disaccharide region of GAG chains. [UniProt].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ACPL2
Polyclonal
Unconjugated
PXYLP1
2-phosphoxylose phosphatase 1; 9430094M07Rik; acid phosphatase-like 2; acid phosphatase-like protein 2; ACPL2; BB177120; C130099A20Rik; epididymis luminal protein 124; HEL124; PXYLP1; PXYLP1 (HGNC:26303); UNQ370/PRO706; Xylosyl phosphatase; XYLP
Rabbit
Antigen Affinity Chromatography
RUO
92370
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q8TE99
PXYLP1
Recombinant protein corresponding to Human ACPL2. Recombinant protein control fragment (Product #RP-90339).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.