missing translation for 'onlineSavingsMsg'
Learn More

ACP-105, MedChemExpress

Product Code. 16493156
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16493156 25 mg 25mg
16454608 10 mg 10mg
16450907 50 mg 50mg
16407327 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16493156 Supplier Medchem Express Supplier No. HY112256

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ACP-105 is an orally available, selective amd potent androgen receptor modulator (SARM), with pEC50s of 9.0 and 9.3 for AR wild type and T877A mutant, respectively.

ACP-105 is an orally available, selective amd potent androgen receptor modulator (SARM), with pEC50s of 9.0 and 9.3 for AR wild type and T877A mutant, respectively.

Specifications

Chemical Name or Material ACP-105
Physical Form Solid
CAS 899821-23-9
Color Light Yellow
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C16H19ClN2O
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : ≥ 103 mg/mL (354.21 mM)
SMILES N#CC1=CC=C(N2[C@@H]3C[C@](O)(C)C[C@H]2CC3)C(C)=C1Cl
Molecular Weight (g/mol) 290.79
Formula Weight 290.79
Percent Purity 99.33%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.