missing translation for 'onlineSavingsMsg'
Learn More

ACP-105, MedChemExpress

Product Code. 16450907
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16450907

Brand: Medchem Express HY112256

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ACP-105 is an orally available, selective amd potent androgen receptor modulator (SARM), with pEC50s of 9.0 and 9.3 for AR wild type and T877A mutant, respectively.

ACP-105 is an orally available, selective amd potent androgen receptor modulator (SARM), with pEC50s of 9.0 and 9.3 for AR wild type and T877A mutant, respectively.

Specifications

Chemical Name or Material ACP-105
Physical Form Solid
CAS 899821-23-9
Color Light Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C16H19ClN2O
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : ≥ 103 mg/mL (354.21 mM)
SMILES N#CC1=CC=C(N2[C@@H]3C[C@](O)(C)C[C@H]2CC3)C(C)=C1Cl
Molecular Weight (g/mol) 290.79
Formula Weight 290.79
Percent Purity 99.33%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.