missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACOX2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA546797
This item is not returnable.
View return policy
Description
Peptide sequence: DQCLNSALGC YDGNVYERLF QWAQKSPTNT QENPAYEEYI RPLLQSWRSK.
The product of this gene belongs to the acyl-CoA oxidase family. It encodes the branched-chain acyl-CoA oxidase which is involved in the degradation of long branched fatty acids and bile acid intermediates in peroxisomes. Deficiency of this enzyme results in the accumulation of branched fatty acids and bile acid intermediates, and may lead to Zellweger syndrome, severe mental retardation, and death in children.
Specifications
| ACOX2 | |
| Polyclonal | |
| Unconjugated | |
| ACOX2 | |
| 3-alpha,7-alpha,12-alpha-trihydroxy-5-beta-cholestanoyl-CoA 24-hydroxylase; 3-alpha,7-alpha,12-alpha-trihydroxy-5-beta-cholestanoyl-CoA oxidase; Acox2; acyl-CoA oxidase 2; acyl-CoA oxidase 2, branched chain; acyl-Coenzyme A oxidase 2, branched chain; BCOX; branched chain acyl-CoA oxidase; BRCACOX; BRCOX; Peroxisomal acyl-coenzyme A oxidase 2; peroxisomal branched chain acyl-CoA oxidase; THCA-CoA oxidase; THCCox; trihydroxycoprostanoyl-CoA oxidase | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 8309 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q99424 | |
| ACOX2 | |
| Synthetic peptide directed towards the C-terminal of human ACOX2 (aa 631-680). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction