Learn More
ACOX1 Antibody (003), mFluor Violet 610 SE, Novus Biologicals™
Product Code. 18404099 Shop All Bio Techne Products
Product Code. 18404099 Supplier Novus Biologicals Supplier No. NBP289864MFV610
Rabbit Monoclonal Antibody
ACOX1 Monoclonal antibody specifically detects ACOX1 in Human samples. It is validated for Immunohistochemistry (Paraffin)