Learn More
Invitrogen™ ACOT8 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588959
Description
Immunogen sequence: MSSPQAPEDG QGCGDRGDPP GDLRSVLVTT VLNLEPLDED LFRGRHYWVP AKRLFGGQIV GQALVAAAKS VSEDVHVHSL HCYFVRAGDP KLPVLYQVER TRTGSSFSVR SVKAVQHGKP IFICQASFQQ AQPSPMQHQF SMPTVPPPEE LLDCETLIDQ YLRDPNLQKR YPLALNRIAA QEVPIEIKPV NPSPLSQLQR MEPKQMFWVR ARGYIGEGDM; Positive Samples: Jurkat, Raji, U-87MG, HeLa; Cellular Location: Peroxisome.
Acyl-CoA thioesterases are a group of enzymes that catalyze the hydrolysis of acyl-CoAs to the free fatty acid and coenzyme A (CoASH), providing the potential to regulate intracellular levels of acyl-CoAs, free fatty acids and CoASH. It may mediate Nef-induced down-regulation of CD4. It may be involved in the metabolic regulation of peroxisome proliferation.
Specifications
| ACOT8 | |
| Polyclonal | |
| Unconjugated | |
| ACOT8 | |
| 4,8-dimethylnonanoyl-CoA thioesterase; 48-dimethylnonanoyl-CoA thioesterase; ACOT8; ACTEIII; Acyl-CoA thioesterase 8; acyl-coenzyme A thioesterase 8; choloyl-CoA hydrolase; choloyl-coenzyme A thioesterase; hACTE-III; HIV-Nef associated acyl-CoA thioesterase; HIV-Nef-associated acyl-CoA thioesterase; HNAACTE; hTE; long-chain fatty-acyl-CoA hydrolase; NAP1; Nef (lentivirus myristoylated factor) associated protein 1; palmitoyl-CoA hydrolase; peroxisomal acyl-CoA thioesterase 1; peroxisomal acyl-CoA thioesterase 2; Peroxisomal acyl-coenzyme A thioester hydrolase 1; peroxisomal long-chain acyl-CoA thioesterase 1; PTE1; PTE-1; PTE2; PTE-2; Thioesterase II; thioesterase III | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10005 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.67 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| O14734 | |
| ACOT8 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human ACOT8 (NP_0054602). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.