missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACOT8 Mouse anti-Human, Clone: 4D10, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59419-100UL
This item is not returnable.
View return policy
Description
ACOT8 Monoclonal specifically detects ACOT8 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| ACOT8 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Flow Cytometry 1:10-1:1029, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
| ACTEIII, acyl-CoA thioesterase 8HNAACTE, Choloyl-coenzyme A thioesterase, EC 3.1.2.27, hACTEIII, HIV-Nef-associated acyl-CoA thioesterase, hTEcholoyl-CoA hydrolase, long-chain fatty-acyl-CoA hydrolase, palmitoyl-CoA hydrolase, peroxisomal acyl-CoA thioesterase, peroxisomal acyl-CoA thioesterase 1, Peroxisomal acyl-coenzyme A thioester hydrolase 1, Peroxisomal long-chain acyl-CoA thioesterase 1, PTE-1, PTE1HIV-Nef associated acyl-CoA thioesterase, PTE2, PTE-2hACTE-IIIacyl-coenzyme A thioesterase 8, Thioesterase II, thioesterase III | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 10005 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| 4D10 | |
| Unconjugated | |
| Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
| ACOT8 | |
| Recombinant human ACOT8 (1-319aa) purified from E. coli | |
| 100 μL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction