missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACOT8 Antibody (4D10), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59419PCP
This item is not returnable.
View return policy
Description
ACOT8 Monoclonal antibody specifically detects ACOT8 in Human samples. It is validated for Flow Cytometry
Specifications
| ACOT8 | |
| Monoclonal | |
| PerCP | |
| ACTEIII, acyl-CoA thioesterase 8HNAACTE, Choloyl-coenzyme A thioesterase, EC 3.1.2.27, hACTEIII, HIV-Nef-associated acyl-CoA thioesterase, hTEcholoyl-CoA hydrolase, long-chain fatty-acyl-CoA hydrolase, palmitoyl-CoA hydrolase, peroxisomal acyl-CoA thioesterase, peroxisomal acyl-CoA thioesterase 1, Peroxisomal acyl-coenzyme A thioester hydrolase 1, Peroxisomal long-chain acyl-CoA thioesterase 1, PTE-1, PTE1HIV-Nef associated acyl-CoA thioesterase, PTE2, PTE-2hACTE-IIIacyl-coenzyme A thioesterase 8, Thioesterase II, thioesterase III | |
| Recombinant human ACOT8 (1-319aa) purified from E. coli | |
| 0.1 mL | |
| Cancer, Cardiovascular Biology, Endocrinology, Immunology, Signal Transduction | |
| 10005 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Flow Cytometry | |
| 4D10 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction