missing translation for 'onlineSavingsMsg'
Learn More

ACOT8 Antibody (4D10), mFluor Violet 610 SE, Novus Biologicals™

Product Code. 18427582 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18427582 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18427582 Supplier Novus Biologicals Supplier No. NBP259419MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ACOT8 Monoclonal antibody specifically detects ACOT8 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACOT8
Applications Flow Cytometry
Classification Monoclonal
Clone 4D10
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias ACTEIII, acyl-CoA thioesterase 8HNAACTE, Choloyl-coenzyme A thioesterase, EC 3.1.2.27, hACTEIII, HIV-Nef-associated acyl-CoA thioesterase, hTEcholoyl-CoA hydrolase, long-chain fatty-acyl-CoA hydrolase, palmitoyl-CoA hydrolase, peroxisomal acyl-CoA thioesterase, peroxisomal acyl-CoA thioesterase 1, Peroxisomal acyl-coenzyme A thioester hydrolase 1, Peroxisomal long-chain acyl-CoA thioesterase 1, PTE-1, PTE1HIV-Nef associated acyl-CoA thioesterase, PTE2, PTE-2hACTE-IIIacyl-coenzyme A thioesterase 8, Thioesterase II, thioesterase III
Host Species Mouse
Immunogen Recombinant human ACOT8 (1-319aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology, Endocrinology, Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10005
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.