missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACOT7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15972-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the acyl coenzyme family. The encoded protein hydrolyzes the CoA thioester of palmitoyl-CoA and other long-chain fatty acids. Decreased expression of this gene may be associated with mesial temporal lobe epilepsy. Alternatively spliced transcript variants encoding distinct isoforms with different subcellular locations have been characterized.Specifications
| ACOT7 | |
| Polyclonal | |
| Unconjugated | |
| ACOT7 | |
| ACH1, ACOT7, ACT, acyl CoA thioesterase 7, BACH, Brain acyl CoA hydrolase, CTE II, CTE IIa, hBACH, LACH, LACH1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11332, 26759, 70025 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O00154, Q64559, Q91V12 | |
| Acot7 | |
| ACOT7 Fusion Protein Ag8759 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction