missing translation for 'onlineSavingsMsg'
Learn More

ACOT7 Antibody (1E7), Novus Biologicals™

Product Code. 18399329 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18399329 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18399329 Supplier Novus Biologicals Supplier No. H00011332M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ACOT7 Monoclonal antibody specifically detects ACOT7 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACOT7
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Sandwich ELISA
Classification Monoclonal
Clone 1E7
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH17365
Gene Alias ACH1, acyl-CoA thioesterase 7CTE-IIa, BACHEC 3.1.2.2, brain acyl CoA hydrolase, Brain acyl-CoA hydrolase, CTE-IILACH, cytosolic acyl coenzyme A thioester hydrolase, hBACH, long chain, Long chain acyl-CoA thioester hydrolase
Host Species Mouse
Immunogen ACOT7 (AAH17365, 1 a.a. ~ 338 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MSGPDVETPSAIQICRIMRPDDANVAGNVHGGTILKMIEEAGAIISTRHCNSQNGERCVAALARVERTDFLSPMCIGEVAHVSAEITYTSKHSVEVQVNVMSENILTGAKKLTNKATLWYVPLSLKNVDKVLEVPPVVYSRQEQEEEGRKRYEAQKLERMETKWRNGDIVQPVLNPEPNTVSYSQSSLIHLVGPSDCTLHGFVHGGVTMKLMDEVAGIVAARHCKTNIVTASVDAINFHDKIRKGCVITISGRMTFTSNKSMEIEVLVDADPVVDSSQKRYRAASAFFTYVSLSQEGRSLPVPQLVPETEDEKKRFEEGKGRYLQMKAKRQGHAEPQP
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 11332
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.