missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACOT4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£342.00 - £470.00
Specifications
| Antigen | ACOT4 |
|---|---|
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18437090
|
Novus Biologicals
NBP1-83539-25ul |
25 μL |
£342.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18292086
|
Novus Biologicals
NBP1-83539 |
0.1 mL |
£470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ACOT4 Polyclonal specifically detects ACOT4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| ACOT4 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| acyl-CoA thioesterase 4EC 3.1.2.2, acyl-coenzyme A thioesterase 4, Peroxisomal acyl coenzyme A thioester hydrolase Ib, peroxisomal acyl-CoA thioesterase 2B, Peroxisomal long-chain acyl-CoA thioesterase Ib, PTE-2b, PTE2BPTE1B, PTEIB, PTE-IbFLJ31235 | |
| ACOT4 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q8N9L9 | |
| 122970 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:PPLGYDLRRIKVAFSGLVDIVDIRNALVGGYKNPSMIPIEKAQGPILLIVGQDDHNWRSELYAQTVSERLQAHGKEKPQIICYPGTGHYIEPPYFPLCPASLHRLLNKHVIWGGEPRAHSKAQEDAWKQILAFFCKHLGGTQKT | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 46 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title