missing translation for 'onlineSavingsMsg'
Learn More

ACOT2 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18504940 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18504940 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18504940 Supplier Novus Biologicals Supplier No. NBP299168AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ACOT2 Polyclonal antibody specifically detects ACOT2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACOT2
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias acyl-CoA thioesterase 2MTE1, Acyl-coenzyme A thioester hydrolase 2a, acyl-coenzyme A thioesterase 2, mitochondrial, CTE-Ia, EC 3.1.2.2, Long-chain acyl-CoA thioesterase 2, mitochondrial acyl-CoA thioesterase 1, mitochondrial acyl-CoA thioesterase 2, peroxisomal long-chain acyl-coA thioesterase 2, PTE2A, PTE2CTE1A, ZAP128Mte1
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human ACOT2
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10965
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.