missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACOT12 Monoclonal Antibody (OTI1B9)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525428
This item is not returnable.
View return policy
Description
ACOT12 Monoclonal Antibody for Western Blot, IHC (P), Flow
Hydrolyzes acetyl-CoA to acetate and CoA.
Specifications
| ACOT12 | |
| Monoclonal | |
| 0.91 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8WYK0 | |
| ACOT12 | |
| Full length human recombinant protein of ACOT12 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human, Monkey | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI1B9 | |
| Unconjugated | |
| ACOT12 | |
| 1300004O04Rik; 4930449F15Rik; Acetyl-coenzyme A thioesterase; Acot12; Acyl-CoA thioester hydrolase 12; acyl-CoA thioesterase 12; Acyl-coenzyme A thioesterase 12; AV027244; Cach; CACH1; CACH-1; Cytoplasmic acetyl-CoA hydrolase 1; cytosolic acetyl-CoA hydrolase; hCACH-1; mCACH-1; rACH; rCACH-1; STARD15; StAR-related lipid transfer (START) domain containing 15; START domain-containing protein 15; THEAL | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 134526 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction