missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACOT12 Antibody (OTI1A11), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72389AF405
This item is not returnable.
View return policy
Description
ACOT12 Monoclonal specifically detects ACOT12 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications
| ACOT12 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ACOT12 | |
| Full length recombinant protein of human ACOT12 (NP_570123) produced in HEK293T cells. | |
| 0.1 mL | |
| 134526.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| OTI1A11 | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| Acyl-CoA thioester hydrolase 12, acyl-CoA thioesterase 12EC 3.1.2.1, acyl-coenzyme A thioesterase 12, Cach, CACH1, CACH-1, Cytoplasmic acetyl-CoA hydrolase 1, cytosolic acetyl-CoA hydrolase, hCACH-1, StARD15, STARD15MGC105114, StAR-related lipid transfer (START) domain containing 15, START domain-containing protein 15, THEAL | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction