missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACOT11 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103564
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ACOT11.
This gene encodes a member of the acyl-CoA thioesterase family which catalyse the conversion of activated fatty acids to the corresponding non-esterified fatty acid and coenzyme A. Expression of a mouse homolog in brown adipose tissue is induced by low temperatures and repressed by warm temperatures. Higher levels of expression of the mouse homolog has been found in obesity-resistant mice compared with obesity-prone mice, suggesting a role of acyl-CoA thioesterase 11 in obesity. Alternative splicing results in transcript variants.
Specifications
| ACOT11 | |
| Polyclonal | |
| Unconjugated | |
| ACOT11 | |
| 1110020M10Rik; 2010309H15Rik; ACOT11; Acyl-CoA thioester hydrolase 11; acyl-CoA thioesterase 11; Acyl-coenzyme A thioesterase 11; adipose-associated thioesterase; AW060409; Bfit; BFIT1; Brown fat-inducible thioesterase; KIAA0707; mKIAA0707; Palmitoyl-coenzyme A thioesterase; STARD14; StAR-related lipid transfer (START) domain containing 14; START domain containing 14; Thea; Them1; thioeste; thioesterase superfamily member 1; thioesterase, adipose associated | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 100363074, 26027, 329910 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8VHQ9, Q8WXI4 | |
| ACOT11 | |
| A synthesized peptide derived from human ACOT11(Accession Q8WXI4), corresponding to amino acid residues V249-G299. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction