missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACOT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA549967
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total ACOT1 protein.
Acyl-CoA thioesterases are a group of enzymes that catalyze the hydrolysis of acyl-CoAs to the free fatty acid and coenzyme A (CoASH), providing the potential to regulate intracellular levels of acyl-CoAs, free fatty acids and CoASH. Active towards fatty acyl-CoA with chain-lengths of C12-C16. Hunt M.C., Submitted (JUN-2005) to the EMBL/GenBank/DDBJ databases. Li W.B., Submitted (JAN-2003) to the EMBL/GenBank/DDBJ databases.
Specifications
| ACOT1 | |
| Polyclonal | |
| Unconjugated | |
| ACOT1 | |
| ACH2; ACOT1; Acyl-CoA thioesterase 1; acyl-coenzyme A thioesterase 1; CTE1; CTE-1; CTE-I; CTE-Ib; Inducible cytosolic acyl-coenzyme A thioester hydrolase; L CTE-I; LACH2; Long chain acyl-CoA hydrolase; long chain acyl-CoA thioester hydrolase; Palmitoyl-coenzyme A thioesterase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 641371 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q86TX2 | |
| ACOT1 | |
| Synthesized peptide derived from internal of human ACOT1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction