missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACOT1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £251.00
Specifications
| Antigen | ACOT1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/ Immunofluorescence 1:100-1:500, Immunoprecipitation 1-5ul/mg of lysate, Immunohistochemistry-Paraffin 1:200-1:1000 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18619156
|
Novus Biologicals
NBP3-12713-100ul |
100 μg |
£251.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18682168
|
Novus Biologicals
NBP3-12713-50ul |
50 μg |
£151.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ACOT1 Polyclonal antibody specifically detects ACOT1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
| ACOT1 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| ACH2, acyl-CoA thioesterase 1Long chain acyl-CoA thioester hydrolase, acyl-coenzyme A thioesterase 1, CTE1, CTE-1, CTE-I, CTE-Ib, EC 3.1.2.2, Inducible cytosolic acyl-coenzyme A thioester hydrolase, LACH2, Long chain acyl-CoA hydrolase | |
| Produced in rabbits immunized with E. coli-derived Human ACOT1 fragment | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/ Immunofluorescence 1:100-1:500, Immunoprecipitation 1-5ul/mg of lysate, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS, pH7.0 | |
| 641371 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title