missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aconitate hydratase B Mouse anti-E. coli, HRP, Antibody Pair, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Solid phase sandwich ELISA for quantitative determination of E. coli Aconitate hydratase B.
Specifications
Specifications
| Antigen | Aconitate hydratase B |
| Applications | ELISA |
| Classification | Monoclonal |
| Conjugate | HRP |
| Gene Alias | acnB, hypothetical protein, yacI, yacJ |
| Host Species | Mouse |
| Quantity | 15 Plates |
| Regulatory Status | RUO |
| Target Species | E. coli |
| Content And Storage | Storage of components varies. See protocol for specific instructions. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?