missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aconitase 2 Antibody (8H4), Novus Biologicals™
Rabbit Monoclonal Antibody
£260.00 - £406.00
Specifications
| Antigen | Aconitase 2 |
|---|---|
| Clone | 8H4 |
| Dilution | ELISA, Immunohistochemistry 1:50-1:200 |
| Applications | ELISA, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043898
|
Novus Biologicals
NBP3-26528-50ul |
50 μL |
£260.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043985
|
Novus Biologicals
NBP3-26528-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Aconitase 2 Monoclonal antibody specifically detects Aconitase 2 in Human samples. It is validated for ELISA, ImmunohistochemistrySpecifications
| Aconitase 2 | |
| ELISA, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Aconitase, aconitase 2, mitochondrial, aconitate hydratase, mitochondrial, ACONM, Citrate hydro-lyase, EC 4.2.1, EC 4.2.1.3, MGC20605, MGC33908 | |
| A synthesized peptide derived from Human Aconitase 2 [UniProt Q99798] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 8H4 | |
| ELISA, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 50 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title